KEY432310538 Key Organics / BIONET

3-[1-(4-methoxyphenyl)-5-oxopyrrolidine-3-amido]benzoic acid

  • MDL number MFCD06132015
  • Cas Number : 894748-00-6
  • Molecular Weight : 354.362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441463

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3382_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart