KEY432310540 Key Organics / BIONET

ethyl 6-(chloromethyl)-4-(4-ethoxyphenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD14703555
  • Cas Number : 1232790-12-3
  • Molecular Weight : 338.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441465

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3384_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart