KEY432310543 Key Organics / BIONET

3-(2-butoxyphenyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030208
  • Cas Number : 899710-38-4
  • Molecular Weight : 260.293

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441468

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3387_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart