KEY432310544 Key Organics / BIONET

1-(4-ethoxyphenyl)-3-propylthiourea

  • MDL number MFCD03140882
  • Cas Number : 431922-74-6
  • Molecular Weight : 238.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441469

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3388_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart