KEY432310548 Key Organics / BIONET

2-({5-[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}sulfanyl)acetic acid

  • MDL number MFCD19104980
  • Cas Number : 1325306-47-5
  • Molecular Weight : 343.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ({5-[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]-2-pyridinyl}sulfanyl)acetic acid
Inchi Code 1S/C16H13N3O4S/c1-22-12-5-2-10(3-6-12)15-18-16(23-19-15)11-4-7-13(17-8-11)24-9-14(20)21/h2-8H,9H2,1H3,(H,20,21)
Inchi Key FLFFINICRRLMNP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441473

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3392_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart