KEY432310549 Key Organics / BIONET

1-[3-(4-methoxyphenyl)-1H-pyrazole-5-carbonyl]piperidin-4-amine

  • MDL number MFCD20528924
  • Molecular Weight : 300.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-{[3-(4-methoxyphenyl)-1H-pyrazol-5-yl]carbonyl}-4-piperidinamine
Inchi Code 1S/C16H20N4O2/c1-22-13-4-2-11(3-5-13)14-10-15(19-18-14)16(21)20-8-6-12(17)7-9-20/h2-5,10,12H,6-9,17H2,1H3,(H,18,19)
Inchi Key SEDFSZUCKVKUIL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441474

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3393_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart