KEY432310551 Key Organics / BIONET

tert-butyl 2-(3,4-dimethylphenyl)-3-sulfanylidene-1,4,8-triazaspiro[4.5]dec-1-ene-8-carboxylate

  • MDL number MFCD14726870
  • Cas Number : 892299-26-2
  • Molecular Weight : 373.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441476

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3395_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart