KEY432310553 Key Organics / BIONET

3-(4-ethoxybenzenesulfonyl)-4-hydroxy-2-oxo-1,2-dihydroquinoline-7-carboxylic acid

  • MDL number MFCD20528974
  • Cas Number : 1351398-45-2
  • Molecular Weight : 389.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441478

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3397_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart