KEY432310555 Key Organics / BIONET

4-hydroxy-3-(4-methylbenzenesulfonyl)-2-oxo-1,2-dihydroquinoline-7-carboxylic acid

  • MDL number MFCD06744909
  • Cas Number : 892271-80-6
  • Molecular Weight : 359.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441480

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3399_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart