KEY432310559 Key Organics / BIONET

3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]pyridine-2-thiol

  • MDL number MFCD19104976
  • Cas Number : 1325304-97-9
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441484

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3403_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart