KEY432310568 Key Organics / BIONET

ethyl 6-(chloromethyl)-4-(3-chlorophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • MDL number MFCD18201575
  • Cas Number : 1260949-64-1
  • Molecular Weight : 329.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441493

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3412_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart