KEY432310583 Key Organics / BIONET

1-chloro-5-fluoro-4-iodo-2-methylbenzene

  • MDL number MFCD16658720
  • Cas Number : 1126424-32-5
  • Molecular Weight : 270.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 0.90
Storage temp. RT
Physical Form Colourless to Orange Low Melting Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441508

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4179_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart