KEY432310592 Key Organics / BIONET

4,4,5,5-tetramethyl-2-[4-(propan-2-yl)phenyl]-1,3,2-dioxaborolane

  • MDL number MFCD15143592
  • Cas Number : 325142-91-4
  • Molecular Weight : 246.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-isopropylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C15H23BO2/c1-11(2)12-7-9-13(10-8-12)16-17-14(3,4)15(5,6)18-16/h7-11H,1-6H3
Inchi Key FURREIFIKVSTMH-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441441

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3098_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart