KEY432310597 Key Organics / BIONET

1,3-dimethoxy-5-[(E)-2-(4-nitrophenyl)ethenyl]benzene

  • MDL number MFCD16038081
  • Cas Number : 586410-18-6
  • Molecular Weight : 285.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 141
Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441522

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KH-0004_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart