KEY432310613 Key Organics / BIONET

S,S-diethyl-sulfoximine

  • MDL number MFCD27933105
  • Cas Number : 92523-32-5
  • Molecular Weight : 121.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441539

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4862_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart