KEY432441772 Key Organics / BIONET

4-[2,2,4-trimethyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1-benzopyran-4-yl]phenol

  • MDL number MFCD28155241
  • Molecular Weight : 394.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441951

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3107_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart