KEY432441773 Key Organics / BIONET

(E)-N'-{5-[(3,4-dimethylcyclohexyl)amino]-4H-1,2,4-triazol-3-yl}-N,N-dimethylmethanimidamide

  • MDL number MFCD28155260
  • Cas Number : 1638617-31-8
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442191

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/JS-2990_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart