KEY432441774 Key Organics / BIONET

ethyl(imino)methyl-lamda(6)-sulfanone

  • MDL number MFCD28155261
  • Cas Number : 35362-83-5
  • Molecular Weight : 107.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442214

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4865_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart