KEY432441777 Key Organics / BIONET

4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulfonyl]morpholine

  • MDL number MFCD14702534
  • Cas Number : 1401222-64-7
  • Molecular Weight : 353.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]sulfonyl}morpholine
Inchi Code 1S/C16H24BNO5S/c1-15(2)16(3,4)23-17(22-15)13-5-7-14(8-6-13)24(19,20)18-9-11-21-12-10-18/h5-8H,9-12H2,1-4H3
Inchi Key ZOTKDMJEHZLGRG-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441946

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2830_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart