KEY432441780 Key Organics / BIONET

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromen-2-one

  • MDL number MFCD27936531
  • Cas Number : 1408287-07-9
  • Molecular Weight : 272.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441954

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3110_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart