KEY432441792 Key Organics / BIONET

ethyl 4-[1-(4-chlorophenyl)-5-oxopyrrolidine-3-amido]benzoate

  • MDL number MFCD00502046
  • Cas Number : 328090-80-8
  • Molecular Weight : 386.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441966

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3430_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart