KEY432441801 Key Organics / BIONET

1,3-diethyl 2-acetamido-2-(2-phenylethyl)propanedioate

  • MDL number MFCD20489108
  • Cas Number : 5463-92-3
  • Molecular Weight : 321.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441975

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3439_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart