KEY432441803 Key Organics / BIONET

(Z)-1-(3-chlorophenyl)-N'-hydroxy-1H-1,2,3-triazole-4-carboximidamide

  • MDL number MFCD17780202
  • Cas Number : 1255791-16-2
  • Molecular Weight : 237.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441977

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3441_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart