KEY432441807 Key Organics / BIONET

3-(3-methylphenyl)-4H,5H-[1,2,3]triazolo[1,5-a]quinazolin-5-one

  • MDL number MFCD06402695
  • Cas Number : 866812-19-3
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441981

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3445_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart