KEY432441808 Key Organics / BIONET

3-(2-methylphenyl)-4H,5H-[1,2,3]triazolo[1,5-a]quinazolin-5-one

  • MDL number MFCD05900312
  • Cas Number : 866808-12-0
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441982

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3446_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart