KEY432441810 Key Organics / BIONET

3-[(4-fluorophenyl)methyl]-1H,2H,3H,4H-thieno[3,2-d]pyrimidine-2,4-dione

  • MDL number MFCD14703392
  • Cas Number : 1232807-82-7
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441984

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3448_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart