KEY432441811 Key Organics / BIONET

2-chloro-N-(2-sulfamoylphenyl)acetamide

  • MDL number MFCD00971884
  • Cas Number : 89981-37-3
  • Molecular Weight : 248.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441985

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3449_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart