KEY432441813 Key Organics / BIONET

(E)-N'-(2,6-dioxo-3-phenyl-1,2,3,6-tetrahydropyrimidin-4-yl)-N,N-dimethylmethanimidamide

  • MDL number MFCD18194591
  • Cas Number : 658061-30-4
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441987

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3451_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart