KEY432441814 Key Organics / BIONET

7-hydroxy-2-(4-methoxyphenyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD22626822
  • Cas Number : 1416340-72-1
  • Molecular Weight : 259.265

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441988

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3452_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart