KEY432441818 Key Organics / BIONET

3-phenyl-1,4-diazaspiro[4.4]non-3-en-2-one

  • MDL number MFCD14735634
  • Cas Number : 899926-21-7
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441992

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3456_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart