KEY432441821 Key Organics / BIONET

ethyl 3-{[(2-hydroxyphenyl)methyl]amino}benzoate

  • MDL number MFCD12462611
  • Cas Number : 1096937-37-9
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-[(2-hydroxybenzyl)amino]benzoate
Inchi Code 1S/C16H17NO3/c1-2-20-16(19)12-7-5-8-14(10-12)17-11-13-6-3-4-9-15(13)18/h3-10,17-18H,2,11H2,1H3
Inchi Key IXDFOUCOLCAQHI-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441995

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3459_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart