KEY432441822 Key Organics / BIONET

ethyl 4-amino-3-(4-nitrophenyl)-2-sulfanylidene-2,3-dihydro-1,3-thiazole-5-carboxylate

  • MDL number MFCD02089352
  • Cas Number : 312922-32-0
  • Molecular Weight : 325.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-amino-3-(4-nitrophenyl)-2-thioxo-2,3-dihydro-1,3-thiazole-5-carboxylate
Inchi Code 1S/C12H11N3O4S2/c1-2-19-11(16)9-10(13)14(12(20)21-9)7-3-5-8(6-4-7)15(17)18/h3-6H,2,13H2,1H3
Inchi Key KBRDKRWIWFVKGJ-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441996

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3460_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart