KEY432441824 Key Organics / BIONET

10-(4-chlorophenyl)-6-methoxy-9-methyl-8-oxa-10,12-diazatricyclo[7.3.1.0^{2,7}]trideca-2,4,6-triene-11-thione

  • MDL number MFCD15086231
  • Cas Number : 702655-76-3
  • Molecular Weight : 360.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/441998

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3462_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart