KEY432441829 Key Organics / BIONET

3,5-diethyl 1-[2-(4-methoxyphenyl)-2-oxoethyl]-1H-pyrazole-3,5-dicarboxylate

  • MDL number MFCD28041455
  • Cas Number : 1638612-94-8
  • Molecular Weight : 360.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442003

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3467_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart