KEY432441830 Key Organics / BIONET

ethyl 5-amino-3-ethyl-1-(2-hydroxy-2-phenylethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD28052833
  • Cas Number : 1638612-77-7
  • Molecular Weight : 303.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442004

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3468_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart