KEY432441831 Key Organics / BIONET

tert-butyl N-{1-[6-(3-fluorophenyl)pyridazin-3-yl]piperidin-4-yl}carbamate

  • MDL number MFCD28041466
  • Cas Number : 1638612-38-0
  • Molecular Weight : 372.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442005

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3469_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart