KEY432441833 Key Organics / BIONET

tert-butyl N-{1-[6-(2-chlorophenyl)pyridazin-3-yl]piperidin-3-yl}carbamate

  • MDL number MFCD28041468
  • Cas Number : 1638612-47-1
  • Molecular Weight : 388.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442007

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3471_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart