KEY432441834 Key Organics / BIONET

6-(3-methylphenyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD02930929
  • Cas Number : 66549-33-5
  • Molecular Weight : 186.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442008

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3472_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart