KEY432441835 Key Organics / BIONET

4-oxo-3-pentyl-2-sulfanylidene-1,2,3,4-tetrahydroquinazoline-7-carboxylic acid

  • MDL number MFCD02111448
  • Cas Number : 309940-73-6
  • Molecular Weight : 292.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442009

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3473_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart