KEY432441837 Key Organics / BIONET

6-benzyl-2-(3-chlorophenyl)-3H,4H,5H,6H,7H,8H-pyrido[4,3-d]pyrimidin-4-one

  • MDL number MFCD28052840
  • Cas Number : 1283274-03-2
  • Molecular Weight : 351.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3475_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart