KEY432441838 Key Organics / BIONET

3-oxo-N-propyl-3,4-dihydroquinoxaline-6-carboxamide

  • MDL number MFCD28041481
  • Cas Number : 1638612-60-8
  • Molecular Weight : 231.255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442012

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3476_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart