KEY432441840 Key Organics / BIONET

3-(pyridin-4-yl)-1H-pyrazole-5-carbohydrazide

  • MDL number MFCD25953250
  • Cas Number : 32725-40-9
  • Molecular Weight : 203.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442014

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3478_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart