KEY432441843 Key Organics / BIONET

bis(N-[(5-methylfuran-2-yl)methyl]guanidine); sulfuric acid

  • MDL number MFCD28155243
  • Cas Number : 30263-43-5
  • Molecular Weight : 404.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3481_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart