KEY432441844 Key Organics / BIONET

2-[3-(4-methoxyphenyl)-1,2,4-oxadiazol-5-yl]acetonitrile

  • MDL number MFCD08741881
  • Cas Number : 892303-79-6
  • Molecular Weight : 215.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442018

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3482_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart