KEY432441845 Key Organics / BIONET

bis(4-(2-fluorophenyl)piperazine-1-carboximidamide); sulfuric acid

  • MDL number MFCD28155244
  • Cas Number : 59083-59-9
  • Molecular Weight : 542.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442019

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3483_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart