KEY432441847 Key Organics / BIONET

bis(N-(2-phenylethyl)guanidine); sulfuric acid

  • MDL number MFCD00035581
  • Cas Number : 2551-72-6
  • Molecular Weight : 424.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442021

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3485_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart