KEY432441848 Key Organics / BIONET

bis(N-[(2-fluorophenyl)methyl]guanidine); sulfuric acid

  • MDL number MFCD28155245
  • Cas Number : 16125-82-9
  • Molecular Weight : 432.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442022

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3486_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart