KEY432441851 Key Organics / BIONET

8-{[(4-methylphenyl)amino]methyl}-2H,3H,6H,7H-[1,4]dioxino[2,3-g]quinolin-7-one

  • MDL number MFCD11048783
  • Cas Number : 947018-69-1
  • Molecular Weight : 322.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442025

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3489_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart