KEY432441853 Key Organics / BIONET

7-{[(4-methoxyphenyl)amino]methyl}-2H,5H,6H-[1,3]dioxolo[4,5-g]quinolin-6-one

  • MDL number MFCD11048792
  • Cas Number : 947019-00-3
  • Molecular Weight : 324.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442027

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3491_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart