KEY432441855 Key Organics / BIONET

methyl 5-{[(4-ethylphenyl)methyl]sulfamoyl}-1H-pyrazole-4-carboxylate

  • MDL number MFCD26130030
  • Molecular Weight : 323.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/442029

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3493_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart